missing translation for 'onlineSavingsMsg'
Learn More

2-Butoxyethyl acetate, 98%, Thermo Scientific Chemicals

Product Code. 11924921
Click to view available options
Quantity:
1 L
2.5 L
10 L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-07-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.21
MDL Number MFCD00009457
InChI Key NQBXSWAWVZHKBZ-UHFFFAOYSA-N
Synonym butoxyethyl acetate, butyl glycol acetate, butylglycol acetate, ethylene glycol monobutyl ether acetate, 2-butoxyethanol acetate, butyl cellosolve acetate, ektasolve eb acetate, ethanol, 2-butoxy-, acetate, butylcelosolvacetat, 1-acetoxy-2-butoxyethane
PubChem CID 8160
IUPAC Name 2-butoxyethyl acetate
SMILES CCCCOCCOC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -64.0°C
Density 0.9385g/mL
Boiling Point 193.5°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic drum
Linear Formula CH3CO2CH2CH2O(CH2)3CH3
Refractive Index 1.4120 to 1.4140
Quantity 10 L
Specific Gravity 0.9385
Solubility Information Solubility in water: soluble
Formula Weight 160.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Butoxyethyl acetate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Butoxyethyl acetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'