missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl-1H-imidazol-4-yl)methanol, 97%, Thermo Scientific™

£10.40 - £109.00

Chemical Identifiers

CAS 68283-19-2
Molecular Formula C8H14N2O
Molecular Weight (g/mol) 154.21
MDL Number MFCD00239516
InChI Key UZKBZGAMRJRWLR-UHFFFAOYSA-N
Synonym 2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
PubChem CID 2735673
IUPAC Name (2-butyl-1H-imidazol-5-yl)methanol
SMILES CCCCC1=NC=C(CO)N1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10029481
View Documents Promotion Details
Thermo Scientific Maybridge
SB02033DA
1 g
£10.40
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10039491
View Documents Promotion Details
Thermo Scientific Maybridge
SB02033EA
10 g
£56.40
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10082404
View Documents Promotion Details
Thermo Scientific Maybridge
SB02033EB
25 g
£109.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

68283-19-2
154.21
UZKBZGAMRJRWLR-UHFFFAOYSA-N
2735673
CCCCC1=NC=C(CO)N1
C8H14N2O
MFCD00239516
2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
(2-butyl-1H-imidazol-5-yl)methanol

Specifications

68283-19-2
97%
C8H14N2O
2-butyl-1h-imidazol-4-yl methanol, 2-butyl-5-hydroxymethylimidazole, 2-butyl-1h-imidazol-5-yl methanol, unii-k14mm5ob9a, 2-butyl-4-hydroxymethyl imidazole, k14mm5ob9a, 2-butylimidazol-5-yl methan-1-ol, 2-butyl-3h-imidazol-4-yl methanol, 2-n-butyl-4-imidazolemethanol, 2-n-butyl-4-hydroxymethylimidazole
CCCCC1=NC=C(CO)N1
154.21
154.21
(2-Butyl-1H-imidazol-4-yl)methanol
97.0
Amber Glass Bottle
MFCD00239516
UZKBZGAMRJRWLR-UHFFFAOYSA-N
(2-butyl-1H-imidazol-5-yl)methanol
2735673
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications