missing translation for 'onlineSavingsMsg'
Learn More

2-Butyne-1,4-diol, 98+%, Thermo Scientific Chemicals

£19.50 - £170.85

Chemical Identifiers

CAS 110-65-6
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002915
InChI Key DLDJFQGPPSQZKI-UHFFFAOYSA-N
Synonym 2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
PubChem CID 8066
ChEBI CHEBI:16413
IUPAC Name but-2-yne-1,4-diol
SMILES C(C#CCO)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11471908
View Documents
Thermo Scientific Alfa Aesar
A14539.30
250 g
£19.50
250g
Please to purchase this item. Need a web account? Register with us today!
11461908
View Documents
Thermo Scientific Alfa Aesar
A14539.0B
1000 g
£44.22
1000g
Please to purchase this item. Need a web account? Register with us today!
11481908
View Documents
Thermo Scientific Alfa Aesar
A14539.0I
5000 g
£170.85
5000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Butyne-1,4-diol is used as a precursor to prepare 1,4-butanediol, 2-butene-1,4-diol and mucochloric acid. It is also used in textile additives, corrosion inhibitors, plasticizers, synthetic resins and polyurethanes. It is an important raw material of vitamin B6. Further, it is used for brightening, preserving and inhibiting nickel plating. In addition, it is used in biological studies for nematocidal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyne-1,4-diol is used as a precursor to prepare 1,4-butanediol, 2-butene-1,4-diol and mucochloric acid. It is also used in textile additives, corrosion inhibitors, plasticizers, synthetic resins and polyurethanes. It is an important raw material of vitamin B6. Further, it is used for brightening, preserving and inhibiting nickel plating. In addition, it is used in biological studies for nematocidal activity.

Solubility
Soluble in water, ethanol, acetone, methanol. Slightly soluble in ethyl ether and chloroform. Insoluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides, strong acids and strong bases.
Specifications

Chemical Identifiers

110-65-6
86.09
DLDJFQGPPSQZKI-UHFFFAOYSA-N
8066
but-2-yne-1,4-diol
C4H6O2
MFCD00002915
2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
CHEBI:16413
C(C#CCO)O

Specifications

110-65-6
194°C (100 mmHg)
C4H6O2
MFCD00002915
1071237
Soluble in water,ethanol,acetone,methanol. Slightly soluble in ethyl ether and chloroform. Insoluble in benzene.
C(C#CCO)O
86.09
CHEBI:16413
≥98%
54°C to 58°C
152°C (305°F)
HOCH2C≡CCH2OH
UN2716
2-butyne-1,4-diol, butynediol, 1,4-dihydroxy-2-butyne, 1,4-butynediol, bis hydroxymethyl acetylene, 2-butynediol, 2-butin-1,4-diol, agrisynth b3d, unii-axh202fpqm, 1,4-butynediol van
DLDJFQGPPSQZKI-UHFFFAOYSA-N
but-2-yne-1,4-diol
8066
86.09
2-Butyne-1,4-diol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H314-H373-H312-H317
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
May cause an allergic skin reaction.

P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H301+H311+H331-H314-H317-H335-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1,4-BUTYNEDIOL

missing translation for 'einecsNumber' : 203-788-6

missing translation for 'rtecsNumber' : ES0525000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt