missing translation for 'onlineSavingsMsg'
Learn More

(2-Carboxyethyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Product Code. 10406832
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51114-94-4
Molecular Formula C21H20BrO2P
Molecular Weight (g/mol) 415.27
MDL Number MFCD00031698
InChI Key BVKRDNIULHRLCO-UHFFFAOYSA-N
Synonym 2-carboxyethyl triphenylphosphonium bromide, 2-carboxyethyl triphenylphosponium bromide, phosphonium, 2-carboxyethyl triphenyl-, bromide, 2-carboxyethyl triphenylphosphanium bromide, c21h20o2p.br, 2-carboxyethyl-triphenylphosphonium bromide, 2-carboxyethyl triphenyl phosphanium bromide, 2-carboxyethyl triphenyl phosphonium bromide
PubChem CID 2733850
IUPAC Name 2-carboxyethyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 415.26
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material (2-Carboxyethyl)triphenylphosphonium bromide, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'