missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate, 96%, Thermo Scientific™

Product Code. 11306435
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy
Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153433-26-2
Molecular Formula C5H10BClF4N2
Molecular Weight (g/mol) 220.40
MDL Number MFCD09039291
InChI Key UPLXKEIRISBKRM-UHFFFAOYSA-N
Synonym CIB
IUPAC Name 2-chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CN1CC[N+](C)=C1Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 58.0°C to 60.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 220.41
Percent Purity 96%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolidinium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-1,3-Dimethylimidazolidinium Tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'