missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 163.58 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-1, 3-thiazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'