missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-tritylimidazole, 97%, Thermo Scientific Chemicals

Product Code. 15418347
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67478-48-2
Molecular Formula C22H17ClN2
Molecular Weight (g/mol) 344.842
MDL Number MFCD02179527
InChI Key JTMBWVKMAGHXKI-UHFFFAOYSA-N
Synonym 2-chloro-1-trityl-1h-imidazole, 2-chloro-1-triphenylmethyl imidazole, pubchem8989, acmc-209nza, 2-chloro-1-trityl-imidazole, 1h-imidazole,2-chloro-1-triphenylmethyl
PubChem CID 2773337
IUPAC Name 2-chloro-1-tritylimidazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=CN=C4Cl

Specifications

Melting Point 200°C to 202°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 344.84
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-tritylimidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'