missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-2-methylnonane, 90+%, Thermo Scientific Chemicals Product Code.: 11300428

2-Chloro-2-methylnonane, 90+%, Thermo Scientific Chemicals

Product Code. 11300428
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11300428

Brand: Thermo Scientific Alfa Aesar 043784.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the photoelectron spectroscopy of C60Fn- and C60Fm2 in the gas phase and the generation and characterization of C1-C60F47- and D2-C60F44 in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4325-50-2
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.728
MDL Number MFCD00060781
InChI Key QDJXQCKHBZEIJM-UHFFFAOYSA-N
Synonym dimethyloctyl chloride, acmc-20ao0l, 2-chloro-2-methyl-nonane, nonane,2-chloro-2-methyl
PubChem CID 138189
IUPAC Name 2-chloro-2-methylnonane
SMILES CCCCCCCC(C)(C)Cl

Specifications

Color Colorless
Quantity 5 g
UN Number UN3082
Formula Weight 176.73
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylnonane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB16L07 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number043784.06,043784.14
Lot NumberH06N30 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number043784.06,043784.14
Lot NumberB18Z015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number043784.06,043784.14
Lot NumberZ08D047 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number043784.06,043784.14
Lot NumberJ06N21 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number043784.06,043784.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-2-methylnonane, 90+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.