missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-methylpropane, 99%

Product Code. 10010590 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10010590

Brand: Thermo Scientific Acros 109500025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

2-Chloro-2-methylpropane, 99%, C4H9Cl, CAS Number-507-20-0, 2-methyl-2-chloropropane, propane, 2-chloro-2-methyl, 2-chloroisobutane, tert-butylchloride, t-butylchloride, chlorotrimethylmethane, trimethylchloromethane, tert-butyl chloride, t-butyl chloride, 2-methyl-2-propyl chloride, 500mL | CAS: 507-20-0 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -25.0°C
Density 0.8500g/mL
Boiling Point 51.0°C to 52.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCl
Refractive Index 1.3838 to 1.3858
Quantity 2.5 L
Beilstein 01, 125
Fieser 02,46
Merck Index 15, 1563
Specific Gravity 0.85
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Viscosity 0.51 mPa.s (20°C)
Formula Weight 92.57
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylpropane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-2-methylpropane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.