Learn More
2-Chloro-3,5-dimethylpyrazine, 97%, Thermo Scientific™
£51.80 - £369.00
Chemical Identifiers
CAS | 38557-72-1 |
---|---|
Molecular Formula | C6H7ClN2 |
Molecular Weight (g/mol) | 142.59 |
MDL Number | MFCD00126945 |
InChI Key | BTGGHNHGPURMEO-UHFFFAOYSA-N |
PubChem CID | 11672680 |
IUPAC Name | 2-chloro-3,5-dimethylpyrazine |
SMILES | CC1=CN=C(Cl)C(C)=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10397663
|
Thermo Scientific Maybridge
MO08011DA |
1 g |
£51.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10153753
|
Thermo Scientific Maybridge
|
5 g |
£206.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10613235
|
Thermo Scientific Maybridge
MO08011EA |
10 g |
£369.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
38557-72-1 | |
142.59 | |
BTGGHNHGPURMEO-UHFFFAOYSA-N | |
2-chloro-3,5-dimethylpyrazine |
C6H7ClN2 | |
MFCD00126945 | |
11672680 | |
CC1=CN=C(Cl)C(C)=N1 |
Specifications
38557-72-1 | |
97% | |
C6H7ClN2 | |
2-chloro 3,5-dimethyl pyarazine, 2-chloro-3,5-dimethyl-1,4-diazine, 2-chloro-3,5-dimethyl-pyrazine, 3,5-dimethyl-2-chloropyrazine, 2-chloro 3,5-dimethyl pyrazine, 2-chloro-3,5-dimethyl pyrazine, pyrazine,2-chloro-3,5-dimethyl, 2-chloranyl-3,5-dimethyl-pyrazine, pyrazine, 2-chloro-3,5-dimethyl, 3-chloro-2,6-dimethylpyrazine | |
CC1=CN=C(Cl)C(C)=N1 | |
142.59 | |
142.59 | |
2-Chloro-3, 5-dimethylpyrazine |
97.0 | |
Amber glass bottle | |
MFCD00126945 | |
BTGGHNHGPURMEO-UHFFFAOYSA-N | |
2-chloro-3,5-dimethylpyrazine | |
11672680 | |
97% |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning