missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)phenol, 98%, Thermo Scientific™

Product Code. 11314246
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138377-34-1
Molecular Formula C7H4ClF3O
Molecular Weight (g/mol) 196.553
MDL Number MFCD00213793
InChI Key BGXZMBZFGAIBDH-UHFFFAOYSA-N
Synonym 2-chloro-3-trifluoromethyl phenol, 2-chloro-3-hydroxybenzotrifluoride, 2-chloro-3-trifluoromethyl-phenol, phenol, 2-chloro-3-trifluoromethyl, pubchem2846, 2-chloranyl-3-trifluoromethyl phenol
PubChem CID 2736601
IUPAC Name 2-chloro-3-(trifluoromethyl)phenol
SMILES C1=CC(=C(C(=C1)O)Cl)C(F)(F)F

Specifications

Density 1.479
Boiling Point 160°C
Flash Point 79°C (174°F)
Refractive Index 1.473
Quantity 1 g
UN Number UN3265
Beilstein 9045773
Formula Weight 196.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'