missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(2-furyl)pyrimidine, 97%, Thermo Scientific™

Product Code. 10749395
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10749395

Brand: Thermo Scientific Maybridge MO07908DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 124959-28-0
Molecular Formula C8H5ClN2O
Molecular Weight (g/mol) 180.59
MDL Number MFCD09065030
InChI Key BLFCXYJTVFTNRX-UHFFFAOYSA-N
Synonym 2-chloro-4-2-furyl pyrimidine, 2-chloro-4-furan-2-yl pyrimidine, 2-2-chloropyrimidin-4-yl furan, pyrimidine,2-chloro-4-2-furanyl, 2-chloro-4-furan-2-yl-pyrimidine, acmc-20mr9m, 2-chloro-4-2-furanyl pyrimidine, pyrimidine, 2-chloro-4-2-furanyl
PubChem CID 14745027
IUPAC Name 2-chloro-4-(furan-2-yl)pyrimidine
SMILES ClC1=NC=CC(=N1)C1=CC=CO1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 180.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(2-furyl)pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.