missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-dimethylpyridine-3-carbonitrile, 97%, Thermo Scientific™

Product Code. 10720350
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14237-71-9
Molecular Formula C8H7ClN2
Molecular Weight (g/mol) 166.61
MDL Number MFCD00051676
InChI Key RETJKTAVEQPNMH-UHFFFAOYSA-N
Synonym 2-chloro-4,6-dimethylnicotinonitrile, 2-chloro-3-cyano-4,6-dimethylpyridine, 2-chloro-4,6-dimethyl-nicotinonitrile, 2-chloro-4,6-dimethyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl, acmc-1azio, chlorodimethylnicotinonitrile, ksc496m6f, 2-chloro-4,6-dimethyinicotinonitrile, 4,6-dimethyl-2-chloro-nicotinonitrile
PubChem CID 223261
IUPAC Name 2-chloro-4,6-dimethylpyridine-3-carbonitrile
SMILES CC1=CC(C)=C(C#N)C(Cl)=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 99.5°C to 101.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 166.61
Percent Purity 97%
Chemical Name or Material 2-chloro-4, 6-dimethylpyridine-3-carbonitrile
compliance-icons
Product Identifier
  • 2-Chloro-4,6-dimethylpyridine-3-carbonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'