Learn More
2-Chloro-4-aminopyridine, 97%, Thermo Scientific Chemicals
£37.99 - £316.58
Chemical Identifiers
CAS | 14432-12-3 |
---|---|
Molecular Formula | C5H5ClN2 |
Molecular Weight (g/mol) | 128.56 |
InChI Key | BLBDTBCGPHPIJK-UHFFFAOYSA-N |
Synonym | 4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine |
PubChem CID | 84432 |
IUPAC Name | 2-chloropyridin-4-amine |
SMILES | C1=CN=C(C=C1N)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10594911
|
Thermo Scientific Acros
345450050 |
5 g | Glass bottle |
£37.99
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041871
|
Thermo Scientific Acros
345450250 |
25 g | Glass bottle |
£126.63
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246873
|
Thermo Scientific Acros
345451000 |
100 g | Glass bottle |
£316.58
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14432-12-3 | |
128.56 | |
4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine | |
2-chloropyridin-4-amine |
C5H5ClN2 | |
BLBDTBCGPHPIJK-UHFFFAOYSA-N | |
84432 | |
C1=CN=C(C=C1N)Cl |
Specifications
14432-12-3 | |
100.0 | |
Yellow | |
97% | |
4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine | |
C1=CN=C(C=C1N)Cl | |
128.56 | |
128.56 | |
Crystalline Powder |
96.0 | |
92°C to 94°C | |
Authentic | |
C5H5ClN2 | |
BLBDTBCGPHPIJK-UHFFFAOYSA-N | |
2-chloropyridin-4-amine | |
84432 | |
97% | |
2-Chloro-4-aminopyridine |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 238-403-
- 2-Chloro-4-aminopyridine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only