missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-1-iodobenzene, 97%, Thermo Scientific™

Catalog Number 10271003
Click to view available options
Quantity:
50 g
100 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
101335-11-9
C6H3ClFI
256.44
MFCD00051774
POTCKVPDYXEGSV-UHFFFAOYSA-N
2-chloro-4-fluoroiodobenzene, 1-chloro-5-fluoro-2-iodobenzene, benzene, 2-chloro-4-fluoro-1-iodo, 2-chloro-4-fluoro-1-iodo-benzene, pubchem3434, acmc-1bu0g, ksc494m4h, 4-fluoro-2-chloro-iodobenzene, 2-chloro-4-fluoro-iodobenzene, 2-chloro-4-fluoro-1-iodobenzol
2736559
2-chloro-4-fluoro-1-iodobenzene
FC1=CC=C(I)C(Cl)=C1
This item is not returnable. View return policy
101335-11-9
C6H3ClFI
256.44
MFCD00051774
POTCKVPDYXEGSV-UHFFFAOYSA-N
2-chloro-4-fluoroiodobenzene, 1-chloro-5-fluoro-2-iodobenzene, benzene, 2-chloro-4-fluoro-1-iodo, 2-chloro-4-fluoro-1-iodo-benzene, pubchem3434, acmc-1bu0g, ksc494m4h, 4-fluoro-2-chloro-iodobenzene, 2-chloro-4-fluoro-iodobenzene, 2-chloro-4-fluoro-1-iodobenzol
2736559
2-chloro-4-fluoro-1-iodobenzene
FC1=CC=C(I)C(Cl)=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 256.44
Percent Purity 97%
Chemical Name or Material 2-chloro-4-fluoro-1-iodobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'