missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-fluorobenzyl bromide, 99%, Thermo Scientific™

Product Code. 10350734
Click to view available options
Quantity:
2.5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45767-66-6
MDL Number MFCD00236025
InChI Key GAUUDQVOPUKGJD-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzyl bromide, 1-bromomethyl-2-chloro-4-fluorobenzene, 2-chloro-4-fluorobenzylbromide, alpha-bromo-2-chloro-4-fluorotoluene, benzene, 1-bromomethyl-2-chloro-4-fluoro, 2-choro-4-fluorobenzylbromide, pubchem4893, 1-bromomethyl-2-chloro-4-fluoro-benzene, ksc493m8r, 2-chloro-4-flurobenzyl bromide
PubChem CID 2725062
IUPAC Name 1-(bromomethyl)-2-chloro-4-fluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)CBr

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C7H5BrClF
Linear Formula F(Cl)C6H3CH2Br
Quantity 100 g
Molecular Weight (g/mol) 223.469
Formula Weight 223.47
Percent Purity ≥98.5%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Chloro-4-fluorobenzyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-4-fluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.