missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-iodoaniline, 98%, Thermo Scientific Chemicals

Product Code. 11469622
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42016-93-3
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00025298
InChI Key MYDAOWXYGPEPJT-UHFFFAOYSA-N
Synonym 2-chloro-4-iodo-phenylamine, benzenamine, 2-chloro-4-iodo, 2-chloro-4-iodo-aniline, ccris 9438, zlchem 542, pubchem3811, 2-chloro-4-iodoanilline, 2-chloro-4-iodo aniline, acmc-1am5p, 2-chloro-4-iodophenylamine
PubChem CID 282930
IUPAC Name 2-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C=C1Cl

Specifications

Melting Point 68°C to 72°C
Quantity 100 g
UN Number UN2811
Beilstein 2081106
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-iodoaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.