missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylaniline, 98%, Thermo Scientific Chemicals

Product Code. 11469997
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-65-6
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007666
InChI Key XGYLSRFSXKAYCR-UHFFFAOYSA-N
Synonym 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
PubChem CID 12007
IUPAC Name 2-chloro-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Cl

Specifications

Melting Point 7°C
Density 1.151
Boiling Point 223°C to 225°C
Flash Point 99°C (210°F)
Refractive Index 1.575
Quantity 100 g
UN Number UN3429
Beilstein 774514
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'