missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11388055
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7073-36-1
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.01
MDL Number MFCD00051515
InChI Key KTHNITVDTYAHFF-UHFFFAOYSA-N
PubChem CID 81522
IUPAC Name 2-chloro-4-nitrobenzoyl chloride
SMILES [O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1

Specifications

Boiling Point 165°C (20 mmHg)
Refractive Index 1.595
Quantity 50 g
UN Number UN3265
Beilstein 519794
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 2-chloro-4-nitro, benzoyl chloride,2-chloro-4-nitro, acmc-209ofu, 2-chloro-4-nitro-benzoyl chloride
Formula Weight 220.01
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrobenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'