Learn More
2-Chloro-4-nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
£33.20 - £414.00
Chemical Identifiers
CAS | 7073-36-1 |
---|---|
Molecular Formula | C7H3Cl2NO3 |
Molecular Weight (g/mol) | 220.01 |
MDL Number | MFCD00051515 |
InChI Key | KTHNITVDTYAHFF-UHFFFAOYSA-N |
PubChem CID | 81522 |
IUPAC Name | 2-chloro-4-nitrobenzoyl chloride |
SMILES | [O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378055
|
Thermo Scientific Alfa Aesar
L08246.06 |
5 g |
£33.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388055
|
Thermo Scientific Alfa Aesar
L08246.18 |
50 g |
£134.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398055
|
Thermo Scientific Alfa Aesar
L08246.30 |
250 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7073-36-1 | |
220.01 | |
KTHNITVDTYAHFF-UHFFFAOYSA-N | |
2-chloro-4-nitrobenzoyl chloride |
C7H3Cl2NO3 | |
MFCD00051515 | |
81522 | |
[O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1 |
Specifications
7073-36-1 | |
C7H3Cl2NO3 | |
MFCD00051515 | |
519794 | |
benzoyl chloride, 2-chloro-4-nitro, benzoyl chloride,2-chloro-4-nitro, acmc-209ofu, 2-chloro-4-nitro-benzoyl chloride | |
[O-][N+](=O)C1=CC=C(C(Cl)=O)C(Cl)=C1 | |
220.01 | |
220.01 | |
2-Chloro-4-nitrobenzoyl chloride |
165°C (20 mmHg) | |
1.595 | |
UN3265 | |
Moisture sensitive | |
KTHNITVDTYAHFF-UHFFFAOYSA-N | |
2-chloro-4-nitrobenzoyl chloride | |
81522 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 230-367-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only