Learn More
2-Chloro-4-nitrophenol, 97%, Thermo Scientific Chemicals
£24.20 - £270.00
Chemical Identifiers
CAS | 619-08-9 |
---|---|
Molecular Formula | C6H4ClNO3 |
Molecular Weight (g/mol) | 173.552 |
MDL Number | MFCD00043910 |
InChI Key | BOFRXDMCQRTGII-UHFFFAOYSA-N |
Synonym | 2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908 |
PubChem CID | 12074 |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])Cl)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11447154
|
Thermo Scientific Alfa Aesar
B21561.09 |
10 g |
£24.20
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457154
|
Thermo Scientific Alfa Aesar
B21561.18 |
50 g |
£76.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11467154
|
Thermo Scientific Alfa Aesar
B21561.30 |
250 g |
£270.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-4-nitrophenol, is frequently used as building blocks for dyes, plastics, explosives. It acts as a catalytic agent, petrochemical additive and used in organic synthesis. It can react with sulfuric acid dimethyl ester to produce 2-chloro-4-nitro-anisole. This reaction will need reagents K2CO3 and xylene.
Solubility
Slightly Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Chemical Identifiers
619-08-9 | |
173.552 | |
BOFRXDMCQRTGII-UHFFFAOYSA-N | |
12074 |
C6H4ClNO3 | |
MFCD00043910 | |
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908 | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O |
Specifications
619-08-9 | |
Characteristic | |
MFCD00043910 | |
2-chloro-4-nitrophenol, nitrofungin, phenol, 2-chloro-4-nitro, 2-chloro-4-nitro phenol, nitrofurgin, phenol, chloro-4-nitro, unii-y2v03m9ul8, 2-chloro-4-nitro-phenol, 2-chloro-4nitrophenol, pubchem18908 | |
BOFRXDMCQRTGII-UHFFFAOYSA-N | |
173.552 | |
173.56 | |
2-Chloro-4-nitrophenol |
108°C to 112°C | |
C6H4ClNO3 | |
2046372 | |
Slightly Soluble in water. | |
C1=CC(=C(C=C1[N+](=O)[O-])Cl)O | |
12074 | |
97% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 210-578-8
missing translation for 'rtecsNumber' : SK5075000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only