Learn More
2-Chloro-4-nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 40397-95-3 |
---|---|
Molecular Formula | C7H3ClN2O3 |
Molecular Weight (g/mol) | 198.56 |
MDL Number | MFCD00037063 |
InChI Key | POLDSAUWRJCSOK-UHFFFAOYSA-N |
Synonym | 2-chloro-4-nitrophenyl isocyanate, 2-chloro-4-nitrophenylisocyanate, acmc-1ama4, 2-chloro-4-nitrobenzenisocyanate, 2-chloro-1-isocyanato-4-nitro-benzene, 2-chloranyl-1-isocyanato-4-nitro-benzene |
PubChem CID | 4614221 |
IUPAC Name | 2-chloro-1-isocyanato-4-nitrobenzene |
SMILES | [O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40397-95-3 | |
198.56 | |
POLDSAUWRJCSOK-UHFFFAOYSA-N | |
4614221 | |
[O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=O |
C7H3ClN2O3 | |
MFCD00037063 | |
2-chloro-4-nitrophenyl isocyanate, 2-chloro-4-nitrophenylisocyanate, acmc-1ama4, 2-chloro-4-nitrobenzenisocyanate, 2-chloro-1-isocyanato-4-nitro-benzene, 2-chloranyl-1-isocyanato-4-nitro-benzene | |
2-chloro-1-isocyanato-4-nitrobenzene |
Specifications
40397-95-3 | |
C7H3ClN2O3 | |
UN2811 | |
Moisture sensitive | |
POLDSAUWRJCSOK-UHFFFAOYSA-N | |
2-chloro-1-isocyanato-4-nitrobenzene | |
4614221 | |
97% |
59°C to 63°C | |
MFCD00037063 | |
2841619 | |
2-chloro-4-nitrophenyl isocyanate, 2-chloro-4-nitrophenylisocyanate, acmc-1ama4, 2-chloro-4-nitrobenzenisocyanate, 2-chloro-1-isocyanato-4-nitro-benzene, 2-chloranyl-1-isocyanato-4-nitro-benzene | |
[O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=O | |
198.56 | |
198.57 | |
2-Chloro-4-nitrophenyl isocyanate |
Safety and Handling
GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
H302+H312+H332-H315-H317-H319-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only