missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10754013
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-54-6
Molecular Formula C7H3Cl2F3O2S
Molecular Weight (g/mol) 279.05
MDL Number MFCD00052912
InChI Key NJXDBSSSDPOAFI-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl benzenesulphonylchloride, 2-chloro-4-trifluoromethyl benzenesulphonyl chloride, 2-chloro-4-trifluoromethyl benzenesulfonylchloride, 2-chloro-4-trifluoromethyl benzenesulfonyl, 2-chloro-4-trifluoromethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 2-chloro-4-trifluoromethyl, pubchem5104, acmc-1bwmt
PubChem CID 2736675
SMILES FC(F)(F)C1=CC(Cl)=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride
Formula Weight 279.06
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'