Learn More
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one, 97%, Thermo Scientific Chemicals
Description
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one is a reagent used in phosphorylation and phosphitylation of alcohols and in the formation of H-phosphonates. It is also reactive cyclic phosphitylating reagent involved in fast coupling rates and hydrolytic cleavage more readily compare to that of a acyclic analog.
Solubility
Soluble in acetonitrile, dichloromethane, tetrahydrofuron and dimethylformamide.
Notes
Light and heat sensitive. Incompatible with water and strong oxidizing agents.

Specifications
Specifications
Melting Point | 36°C to 40°C |
Boiling Point | 127°C to 128°C (11 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
UN Number | UN3261 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in acetonitrile,dichloromethane,tetrahydrofuron and dimethylformamide. |
Formula Weight | 202.54 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4H-1,3,2-benzodioxaphosphorin-4-one |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10231900 | Certificate of Analysis | 28/12/2023 | B21086.06,B21086.14 |
10229097 | Certificate of Analysis | 20/07/2023 | B21086.06,B21086.14,B21086 |
10231900 | Certificate of Analysis | 23/11/2021 | B21086.06,B21086.14 |
10215926 | Certificate of Analysis | 23/11/2021 | B21086.06,B21086.14 |
10179065 | Certificate of Analysis | 23/11/2021 | B21086.06,B21086.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.