Learn More
2-Chloro-5-fluorophenol, 98%, Thermo Scientific™
£41.00 - £155.10
Chemical Identifiers
CAS | 3827-49-4 |
---|---|
Molecular Formula | C6H4ClFO |
Molecular Weight (g/mol) | 146.55 |
MDL Number | MFCD00042524 |
InChI Key | CMQOZIKIOASEIN-UHFFFAOYSA-N |
Synonym | 2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231 |
PubChem CID | 2724522 |
IUPAC Name | 2-chloro-5-fluorophenol |
SMILES | OC1=CC(F)=CC=C1Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10358660
|
Thermo Scientific Acros
249960050 |
5 g | Glass bottle |
£41.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10423835
|
Thermo Scientific Acros
249960250 |
25 g | Glass bottle | ||||||
Description
Chemical Identifiers
3827-49-4 | |
146.55 | |
CMQOZIKIOASEIN-UHFFFAOYSA-N | |
2724522 | |
OC1=CC(F)=CC=C1Cl |
C6H4ClFO | |
MFCD00042524 | |
2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231 | |
2-chloro-5-fluorophenol |
Specifications
3827-49-4 | |
100.0 | |
183°C to 185°C | |
97.5% min. (GC) | |
FC6H3(OH)Cl | |
MFCD00042524 | |
Solubility in water: slightly soluble. Other solubilities: soluble in aqueous alkali,soluble in most common organic solvents | |
OC1=CC(F)=CC=C1Cl | |
146.55 | |
146.55 | |
Liquid |
97.5 | |
Colorless to Yellow | |
Authentic | |
C6H4ClFO | |
1.5240 to 1.526 | |
2-chloro-5-fluoro-phenol, pubchem2146, 2-chloro-5-fluorphenol, acmc-209iz2, ksc226k0t, attercop-chm at137231 | |
CMQOZIKIOASEIN-UHFFFAOYSA-N | |
2-chloro-5-fluorophenol | |
2724522 | |
98% | |
2-Chloro-5-fluorophenol |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only