missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxy-3-methylpyridine, 97%, Thermo Scientific™

Product Code. 15432429
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54232-03-0
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD11519295
InChI Key UGDSWVXJJFIVAH-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
PubChem CID 12852015
IUPAC Name 6-chloro-5-methylpyridin-3-ol
SMILES CC1=CC(O)=CN=C1Cl

Specifications

Quantity 1 g
Formula Weight 143.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-hydroxy-3-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'