missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-hydroxy-3-methylpyridine, 97%, Thermo Scientific™

£61.88 - £241.00

Chemical Identifiers

CAS 54232-03-0
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD11519295
InChI Key UGDSWVXJJFIVAH-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
PubChem CID 12852015
IUPAC Name 6-chloro-5-methylpyridin-3-ol
SMILES CC1=CC(O)=CN=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442429
View Documents
Thermo Scientific Alfa Aesar
H64986.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15432429
View Documents
Thermo Scientific Alfa Aesar
H64986.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

54232-03-0
143.57
UGDSWVXJJFIVAH-UHFFFAOYSA-N
12852015
CC1=CC(O)=CN=C1Cl
C6H6ClNO
MFCD11519295
2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
6-chloro-5-methylpyridin-3-ol

Specifications

54232-03-0
MFCD11519295
UGDSWVXJJFIVAH-UHFFFAOYSA-N
6-chloro-5-methylpyridin-3-ol
12852015
97%
C6H6ClNO
2-chloro-5-hydroxy-3-methylpyridine, 2-chloro-5-hydroxy-3-methylpyrdine, 6-chloro-5-methyl-pyridin-3-ol, 3-pyridinol, 6-chloro-5-methyl, 2-chloro-5-hydroxy-3-picoline, 6-chloro-5-methyl-3-pyridinol, 2-chloro-3-methyl-5-hydroxypyridine, pubchem12478, acmc-209len, 6-chloro-5-methylpyridine-3-ol
CC1=CC(O)=CN=C1Cl
143.57
143.57
2-Chloro-5-hydroxy-3-methylpyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications