missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-iodopyrimidine, 95%, Thermo Scientific™
£82.90 - £290.55
Chemical Identifiers
CAS | 32779-38-7 |
---|---|
Molecular Formula | C4H2ClIN2 |
Molecular Weight (g/mol) | 240.43 |
MDL Number | MFCD06200210 |
InChI Key | WSZRCNZXKKTLQE-UHFFFAOYSA-N |
PubChem CID | 1714223 |
IUPAC Name | 2-chloro-5-iodopyrimidine |
SMILES | ClC1=NC=C(I)C=N1 |
Description
Chemical Identifiers
32779-38-7 | |
240.43 | |
WSZRCNZXKKTLQE-UHFFFAOYSA-N | |
2-chloro-5-iodopyrimidine |
C4H2ClIN2 | |
MFCD06200210 | |
1714223 | |
ClC1=NC=C(I)C=N1 |
Specifications
32779-38-7 | |
94% min. (GC) | |
MFCD06200210 | |
WSZRCNZXKKTLQE-UHFFFAOYSA-N | |
2-chloro-5-iodopyrimidine | |
1714223 | |
95% |
135°C to 136°C | |
C4H2ClIN2 | |
pyrimidine, 2-chloro-5-iodo, 2-chloro-5-iodo-pyrimidine, pubchem21488, acmc-209hvo, 2-cloro-5-iodopyrimidine, 5-iodo-2-chloropyrimidine, ksc569e7p, pyrimidine,2-chloro-5-iodo | |
ClC1=NC=C(I)C=N1 | |
240.43 | |
240.43 | |
2-Chloro-5-iodopyrimidine |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Chloro-5-iodopyrimidine, 95%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only