missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-methoxybenzoic acid, 98%, Thermo Scientific™

Product Code. 15453387
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6280-89-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00466261
InChI Key AQHFCRYZABKUEV-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxy-benzoic acid, 6-chloro-m-anisic acid, benzoic acid, 2-chloro-5-methoxy, 2-chloro-5-methoxybenzoicacid, pubchem4563, zlchem 1016, acmc-20a0mp, 3-carboxy-4-chloroanisole, 2-chloro-5-methoxybenzoic acid
PubChem CID 221503
IUPAC Name 2-chloro-5-methoxybenzoic acid
SMILES COC1=CC=C(Cl)C(=C1)C(O)=O

Specifications

Melting Point 171°C to 175°C
Quantity 250 mg
Formula Weight 186.59
Percent Purity 98%
Chemical Name or Material 2-Chloro-5-methoxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'