missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(methylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 15402196
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2096338-17-7
Molecular Formula C8H9BClNO3
Molecular Weight (g/mol) 213.42
MDL Number MFCD22125158
InChI Key YTEOTIIPHSQWBH-UHFFFAOYSA-N
Synonym 2-chloro-5-methylcarbamoyl benzeneboronic acid, 2-chloro-5-methylcarbamoyl phenylboronic acid
PubChem CID 73996153

Specifications

Quantity 1 g
Formula Weight 213.43
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(methylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.