missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-methylpyridine-3-boronic acid, 98%, Thermo Scientific™
£101.49 - £404.29
Chemical Identifiers
CAS | 913835-86-6 |
---|---|
Molecular Formula | C6H7BClNO2 |
Molecular Weight (g/mol) | 171.39 |
InChI Key | ZMHWEBDZKPQNHG-UHFFFAOYSA-N |
Synonym | 2-chloro-5-methylpyridine-3-boronic acid, 2-chloro-5-methylpyridin-3-yl boronic acid, 2-chloro-5-picoline-3-boronic acid, 2-chloro-5-methylpyridine-3-boronicacid, alphachiron 13000b651, boronicacid, b-2-chloro-5-methyl-3-pyridinyl, 2-chloro-5-methyl-3-pyridineboronic acid, pubchem11116, acmc-209ra8, abbypharma ap-16-5787 |
PubChem CID | 2763126 |
IUPAC Name | (2-chloro-5-methylpyridin-3-yl)boronic acid |
SMILES | B(C1=CC(=CN=C1Cl)C)(O)O |
Description
Chemical Identifiers
913835-86-6 | |
171.39 | |
2-chloro-5-methylpyridine-3-boronic acid, 2-chloro-5-methylpyridin-3-yl boronic acid, 2-chloro-5-picoline-3-boronic acid, 2-chloro-5-methylpyridine-3-boronicacid, alphachiron 13000b651, boronicacid, b-2-chloro-5-methyl-3-pyridinyl, 2-chloro-5-methyl-3-pyridineboronic acid, pubchem11116, acmc-209ra8, abbypharma ap-16-5787 | |
(2-chloro-5-methylpyridin-3-yl)boronic acid |
C6H7BClNO2 | |
ZMHWEBDZKPQNHG-UHFFFAOYSA-N | |
2763126 | |
B(C1=CC(=CN=C1Cl)C)(O)O |
Specifications
913835-86-6 | |
Authentic | |
C6H7BClNO2 | |
ZMHWEBDZKPQNHG-UHFFFAOYSA-N | |
(2-chloro-5-methylpyridin-3-yl)boronic acid | |
2763126 | |
98% |
115.5°C to 119.0°C | |
97.5% min. (HPLC) | |
2-chloro-5-methylpyridine-3-boronic acid, 2-chloro-5-methylpyridin-3-yl boronic acid, 2-chloro-5-picoline-3-boronic acid, 2-chloro-5-methylpyridine-3-boronicacid, alphachiron 13000b651, boronicacid, b-2-chloro-5-methyl-3-pyridinyl, 2-chloro-5-methyl-3-pyridineboronic acid, pubchem11116, acmc-209ra8, abbypharma ap-16-5787 | |
B(C1=CC(=CN=C1Cl)C)(O)O | |
171.39 | |
171.39 | |
2-Chloro-5-methylpyridine-3-boronic acid |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Chloro-5-methylpyridine-3-boronic acid, 98%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only