missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-n-decylpyrimidine, 99%, Thermo Scientific™

£48.02 - £187.00

Chemical Identifiers

CAS 170434-06-7
Molecular Formula C14H23ClN2
Molecular Weight (g/mol) 254.80
MDL Number MFCD02683077
InChI Key LJRSECCCAXFYFU-UHFFFAOYSA-N
Synonym 2-chloro-5-n-decylpyrimidine
PubChem CID 22099083
IUPAC Name 2-chloro-5-decylpyrimidine
SMILES CCCCCCCCCCC1=CN=C(Cl)N=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11444957
View Documents
Thermo Scientific Alfa Aesar
B21889.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11454957
View Documents
Thermo Scientific Alfa Aesar
B21889.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

170434-06-7
254.80
LJRSECCCAXFYFU-UHFFFAOYSA-N
22099083
CCCCCCCCCCC1=CN=C(Cl)N=C1
C14H23ClN2
MFCD02683077
2-chloro-5-n-decylpyrimidine
2-chloro-5-decylpyrimidine

Specifications

170434-06-7
MFCD02683077
LJRSECCCAXFYFU-UHFFFAOYSA-N
2-chloro-5-decylpyrimidine
22099083
99%
C14H23ClN2
2-chloro-5-n-decylpyrimidine
CCCCCCCCCCC1=CN=C(Cl)N=C1
254.80
254.8
2-Chloro-5-n-decylpyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications