missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-5-nitroaniline, 98%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 117°C to 121°C |
Flash Point | 191°C (375°F) |
Odor | Weak, Characteristic |
Quantity | 100 g |
UN Number | UN2237 |
Beilstein | 2208878 |
Formula Weight | 172.57 |
Percent Purity | 98% |
Chemical Name or Material | 2-Chloro-5-nitroaniline |
Safety and Handling
Product Identifier
- 2-Chloro-5-nitroaniline
- Danger
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after repeated exposure Category 2
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : 2-Chloro-5-nitroaniline
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'