Learn More
2-Chloro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
£20.40 - £50.80
Chemical Identifiers
CAS | 777-37-7 |
---|---|
Molecular Formula | C7H3ClF3NO2 |
Molecular Weight (g/mol) | 225.55 |
MDL Number | MFCD00007296 |
InChI Key | HQROXDLWVGFPDE-UHFFFAOYSA-N |
Synonym | 2-chloro-5-nitrobenzotrifluoride, 1-chloro-4-nitro-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl nitrobenzene, benzene, 1-chloro-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-chloronitrobenzene, 2-trifluoromethyl-4-nitrochlorobenzene, 2-chloro-5-nitro-a,a,a-trifluorotoluene, 4-nitro-2-trifluoromethyl chlorobenzene, 1-chloro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-2-chlorobenzotrifluoride |
PubChem CID | 61213 |
IUPAC Name | 1-chloro-4-nitro-2-(trifluoromethyl)benzene |
SMILES | [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356926
|
Thermo Scientific Alfa Aesar
L06623.14 |
25 g |
£20.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366926
|
Thermo Scientific Alfa Aesar
L06623.22 |
100 g |
£50.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
777-37-7 | |
225.55 | |
HQROXDLWVGFPDE-UHFFFAOYSA-N | |
61213 | |
[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F |
C7H3ClF3NO2 | |
MFCD00007296 | |
2-chloro-5-nitrobenzotrifluoride, 1-chloro-4-nitro-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl nitrobenzene, benzene, 1-chloro-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-chloronitrobenzene, 2-trifluoromethyl-4-nitrochlorobenzene, 2-chloro-5-nitro-a,a,a-trifluorotoluene, 4-nitro-2-trifluoromethyl chlorobenzene, 1-chloro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-2-chlorobenzotrifluoride | |
1-chloro-4-nitro-2-(trifluoromethyl)benzene |
Specifications
777-37-7 | |
1.547 | |
98°C (208°F) | |
1.508 | |
2216169 | |
HQROXDLWVGFPDE-UHFFFAOYSA-N | |
1-chloro-4-nitro-2-(trifluoromethyl)benzene | |
61213 | |
98% |
∼22°C | |
232°C | |
C7H3ClF3NO2 | |
MFCD00007296 | |
2-chloro-5-nitrobenzotrifluoride, 1-chloro-4-nitro-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl nitrobenzene, benzene, 1-chloro-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-chloronitrobenzene, 2-trifluoromethyl-4-nitrochlorobenzene, 2-chloro-5-nitro-a,a,a-trifluorotoluene, 4-nitro-2-trifluoromethyl chlorobenzene, 1-chloro-2-trifluoromethyl-4-nitrobenzene, 5-nitro-2-chlorobenzotrifluoride | |
[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(F)(F)F | |
225.55 | |
225.55 | |
2-Chloro-5-nitrobenzotrifluoride |
Safety and Handling
GHS H Statement
H311-H332-H315-H319
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 212-287-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only