missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-nitrobenzoyl chloride, 96%, Thermo Scientific Chemicals

Product Code. 11308486
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25784-91-2
Molecular Formula C7H3Cl2NO3
Molecular Weight (g/mol) 220.005
MDL Number MFCD00059180
InChI Key OGLKKYALUKXVPQ-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoylchloride, benzoyl chloride, 2-chloro-5-nitro, acmc-1cq6z, 2-chloro-5nitrobenzoyl chloride, 2-chloro-5-nitro-benzoylchlorid, 2-chloro-5-nitro-benzoic chloride, 2-chloro-5-nitro-benzoyl chloride, benzoyl chloride,2-chloro-5-nitro, 2-chloranyl-5-nitro-benzoyl chloride, 2-chloro-5-nitrobenzoic acid chloride
PubChem CID 117637
IUPAC Name 2-chloro-5-nitrobenzoyl chloride
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)Cl)Cl

Specifications

Melting Point 58°C to 60°C
Boiling Point 157°C to 158°C (11 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 2213721
Sensitivity Moisture sensitive
Formula Weight 220.01
Percent Purity 96%
Chemical Name or Material 2-Chloro-5-nitrobenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'