Learn More
2-chloro-5-(trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™
£9.20 - £48.30
Chemical Identifiers
CAS | 54090-08-3 |
---|---|
Molecular Formula | C7H3Cl2F3O2S |
Molecular Weight (g/mol) | 279.054 |
MDL Number | MFCD00052310 |
InChI Key | ZEYKLMDPUOVUCR-UHFFFAOYSA-N |
Synonym | 2-chloro-5-trifluoromethyl benzenesulfonyl chloride, 2-chloro-5-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-5-trifluoromethyl benzenesulphonylchloride, 2-chloro-5-trifluoromethyl benzenesulfonylchloride, chloro 2-chloro-5-trifluoromethyl phenyl sulfone, pubchem5106, acmc-209lcy, 2-chloro-5-trifluoromethylbenzenesulfonylchloride, 2-chloro-5-trifluoromethyl phenylsulfonyl chloride, 4-chloro-3-trifluoromethylbenzene sulfonyl chloride |
PubChem CID | 2734274 |
IUPAC Name | 2-chloro-5-(trifluoromethyl)benzenesulfonyl chloride |
SMILES | C1=CC(=C(C=C1C(F)(F)F)S(=O)(=O)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10032224
|
Thermo Scientific Maybridge
TL00339DA |
1 g | Amber Glass Bottle |
£9.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10555764
|
Thermo Scientific Maybridge
TL00339EA |
10 g | Amber Glass Bottle |
£48.30
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
54090-08-3 | |
279.054 | |
ZEYKLMDPUOVUCR-UHFFFAOYSA-N | |
2734274 | |
C1=CC(=C(C=C1C(F)(F)F)S(=O)(=O)Cl)Cl |
C7H3Cl2F3O2S | |
MFCD00052310 | |
2-chloro-5-trifluoromethyl benzenesulfonyl chloride, 2-chloro-5-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-5-trifluoromethyl benzenesulphonylchloride, 2-chloro-5-trifluoromethyl benzenesulfonylchloride, chloro 2-chloro-5-trifluoromethyl phenyl sulfone, pubchem5106, acmc-209lcy, 2-chloro-5-trifluoromethylbenzenesulfonylchloride, 2-chloro-5-trifluoromethyl phenylsulfonyl chloride, 4-chloro-3-trifluoromethylbenzene sulfonyl chloride | |
2-chloro-5-(trifluoromethyl)benzenesulfonyl chloride |
Specifications
54090-08-3 | |
97% | |
MFCD00052310 | |
ZEYKLMDPUOVUCR-UHFFFAOYSA-N | |
2-chloro-5-(trifluoromethyl)benzenesulfonyl chloride | |
2734274 | |
97% |
97.0 | |
C7H3Cl2F3O2S | |
2-chloro-5-trifluoromethyl benzenesulfonyl chloride, 2-chloro-5-trifluoromethyl benzene-1-sulfonyl chloride, 2-chloro-5-trifluoromethyl benzenesulphonylchloride, 2-chloro-5-trifluoromethyl benzenesulfonylchloride, chloro 2-chloro-5-trifluoromethyl phenyl sulfone, pubchem5106, acmc-209lcy, 2-chloro-5-trifluoromethylbenzenesulfonylchloride, 2-chloro-5-trifluoromethyl phenylsulfonyl chloride, 4-chloro-3-trifluoromethylbenzene sulfonyl chloride | |
C1=CC(=C(C=C1C(F)(F)F)S(=O)(=O)Cl)Cl | |
279.054 | |
279.06 | |
2-chloro-5-(trifluoromethyl)benzene-1-sulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger