Learn More
2-Chloro-5-(trifluoromethyl)pyrazine, 97%, Thermo Scientific Chemicals
£54.70 - £142.00
Chemical Identifiers
CAS | 799557-87-2 |
---|---|
Molecular Formula | C5H2ClF3N2 |
Molecular Weight (g/mol) | 182.53 |
MDL Number | MFCD13189478 |
InChI Key | AIEGIFIEQXZBCP-UHFFFAOYSA-N |
Synonym | 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine |
PubChem CID | 18987645 |
IUPAC Name | 2-chloro-5-(trifluoromethyl)pyrazine |
SMILES | C1=C(N=CC(=N1)Cl)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15456747
|
Thermo Scientific Alfa Aesar
H33863.MD |
250 mg |
£54.70
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446747
|
Thermo Scientific Alfa Aesar
H33863.03 |
1 g |
£142.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
799557-87-2 | |
182.53 | |
AIEGIFIEQXZBCP-UHFFFAOYSA-N | |
18987645 | |
C1=C(N=CC(=N1)Cl)C(F)(F)F |
C5H2ClF3N2 | |
MFCD13189478 | |
2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine | |
2-chloro-5-(trifluoromethyl)pyrazine |
Specifications
799557-87-2 | |
C5H2ClF3N2 | |
UN3267 | |
AIEGIFIEQXZBCP-UHFFFAOYSA-N | |
2-chloro-5-(trifluoromethyl)pyrazine | |
18987645 | |
97% |
85°C (112 mmHg) | |
MFCD13189478 | |
2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine | |
C1=C(N=CC(=N1)Cl)C(F)(F)F | |
182.53 | |
182.53 | |
2-Chloro-5-(trifluoromethyl)pyrazine |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.