missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15488866
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15488866

Brand: Thermo Scientific Alfa Aesar H64361.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It plays an important role for intermediate for pharmaceutical, organic synthesis and organic light-emitting diode(OLED).

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505084-58-2
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD01862019
InChI Key OIODRBHIESSYRO-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyridine-4-carboxylic acid, 2-chloro-5-trifluoromethyl isonicotinic acid, 2-chloro-5-trifluoromethyl-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2-chloro-5-trifluoromethyl, 4-pyridinecarboxylicacid, 2-chloro-5-trifluoromethyl, acmc-209kng, ksc268s9f, 2-chloro-5-trifluoromethylisonicotinic acid, 2-chloro-5-trifluoromethyl isonicotinicacid
PubChem CID 11264608
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
SMILES C1=C(C(=CN=C1Cl)C(F)(F)F)C(=O)O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 225.55
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.