missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-fluoro-3-methylphenol, 97%, Thermo Scientific™

Product Code. 11382689
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261762-90-7
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.572
MDL Number MFCD01631366
InChI Key RPTGCWUNGWVQET-UHFFFAOYSA-N
Synonym acmc-20anyl, 2-chloro-6-fluoro-m-cresol, 2-chloro-6-fluoro-3-methyl-phenol, 2-chloro-4-fluoro-3-hydroxytoluene
PubChem CID 2773683
IUPAC Name 2-chloro-6-fluoro-3-methylphenol
SMILES CC1=C(C(=C(C=C1)F)O)Cl

Specifications

Melting Point 47°C to 50°C
Boiling Point 211°C
Refractive Index 1.505
Quantity 25 g
UN Number UN3261
Formula Weight 160.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-fluoro-3-methylphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'