missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-fluoroaniline, 99%, Thermo Scientific Chemicals

Product Code. 10242802
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-51-9
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00040309
InChI Key ZJLAWMDJTMMTQB-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-6-fluoro, 2-chloro-6-fluoro-aniline, 2-chloro-6-fluoro-phenylamine, 6-chloro-2-fluorophenylamine, pubchem1528, 2-fluoro-6-chloroaniline, acmc-209im1, 2-chloro-6-fluoro aniline, 2-chloro-6-fluorobenzenamine, 2-chloro-6-fluorophenylamine
PubChem CID 2734205
IUPAC Name 2-chloro-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Cl

Specifications

CAS Min % 98.5
Color Yellow
CAS Max % 100.0
Density 1.3160g/mL
Boiling Point 67°C to 69°C (14.0 mmHg)
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5480 to 1.554
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.316
Formula Weight 145.56
Percent Purity 99%
Chemical Name or Material 2-chloro-6-fluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'