missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-fluorobenzene-1-carbonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 193 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-6-fluorobenzene-1-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'