Learn More
2-Chloro-6-fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
£20.50 - £133.00
Chemical Identifiers
CAS | 37777-76-7 |
---|---|
Molecular Formula | C8H6ClFO2 |
Molecular Weight (g/mol) | 188.582 |
MDL Number | MFCD00004319 |
InChI Key | GUAIAAXDEJZRBP-UHFFFAOYSA-N |
Synonym | 2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114 |
PubChem CID | 123464 |
IUPAC Name | 2-(2-chloro-6-fluorophenyl)acetic acid |
SMILES | C1=CC(=C(C(=C1)Cl)CC(=O)O)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
37777-76-7 | |
188.582 | |
GUAIAAXDEJZRBP-UHFFFAOYSA-N | |
123464 | |
C1=CC(=C(C(=C1)Cl)CC(=O)O)F |
C8H6ClFO2 | |
MFCD00004319 | |
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114 | |
2-(2-chloro-6-fluorophenyl)acetic acid |
Specifications
37777-76-7 | |
C8H6ClFO2 | |
4401142 | |
GUAIAAXDEJZRBP-UHFFFAOYSA-N | |
2-(2-chloro-6-fluorophenyl)acetic acid | |
123464 | |
98% |
120°C to 124°C | |
MFCD00004319 | |
2-chloro-6-fluorophenylacetic acid, 2-2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenyl acetic acid, 2-chloro-6-fluorophenylaceticacid, benzeneacetic acid, 2-chloro-6-fluoro, 2-chloro-6-fluoro-phenyl-acetic acid, pubchem4254, acmc-1aewm, rarechem al bo 0114 | |
C1=CC(=C(C(=C1)Cl)CC(=O)O)F | |
188.582 | |
188.59 | |
2-Chloro-6-fluorophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 253-661-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only