missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-hydroxypyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.
Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
6-chloro-2-hydroxypyridine was used to study the synthesis, electrochemistry, spectroscopic properties and X-ray crystal structure of Os2Cl(chp)4 (chp = 6-chloro-2- hydroxypyridinate) complex. It undergoes melt reaction with Os2(O2CCH3)4CI2 at 145°C to yield Os2CI4(chp) and Os2Cl(chp)4.
Notes
Store at room temperature. Ensure adequate ventilation. Incompatible with oxidizing agents.
Specifications
Melting Point | 129°C to 131°C |
Flash Point | 163°C (325°F) |
Quantity | 25 g |
Beilstein | 108665 |
Formula Weight | 129.55 |
Percent Purity | 98% |
Chemical Name or Material | 2-Chloro-6-hydroxypyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'