Learn More
2-Chloro-6-methoxy-3-nitropyridine, 98%, Thermo Scientific Chemicals
£58.80 - £204.00
Chemical Identifiers
CAS | 38533-61-8 |
---|---|
Molecular Formula | C6H5ClN2O3 |
Molecular Weight (g/mol) | 188.57 |
MDL Number | MFCD00130268 |
InChI Key | DVRGUTNVDGIKTP-UHFFFAOYSA-N |
Synonym | 2-chloro-3-nitro-6-methoxypyridine, pyridine, 2-chloro-6-methoxy-3-nitro, 2-chloro-6-methoxy-3-nitro-pyridine, 2-chloro-6-methoxy-3 or 5-nitropyridine, pubchem4044, acmc-1agmz, ksc226c7t, 2-chloro-3nitro-6-methoxy-pyridine, 6-methoxy-2-chloro-3-nitropyridine, 2-chloro-6-methoxy-3-nitropyridine |
PubChem CID | 2795029 |
IUPAC Name | 2-chloro-6-methoxy-3-nitropyridine |
SMILES | COC1=CC=C(C(Cl)=N1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15421119
|
Thermo Scientific Alfa Aesar
H64076.14 |
25 g |
£58.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15431119
|
Thermo Scientific Alfa Aesar
H64076.22 |
100 g |
£204.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
6-methoxy-3-nitropyridine-2-carbonitrile was synthesised from 2-Chloro-6-methoxy-3-nitropyridine. Reatant in the suzuki and negishi couplings reaction. Also as fine chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-methoxy-3-nitropyridine-2-carbonitrile was synthesised from 2-Chloro-6-methoxy-3-nitropyridine. Reatant in the suzuki and negishi couplings reaction. Also as fine chemical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
38533-61-8 | |
188.57 | |
DVRGUTNVDGIKTP-UHFFFAOYSA-N | |
2795029 | |
COC1=CC=C(C(Cl)=N1)[N+]([O-])=O |
C6H5ClN2O3 | |
MFCD00130268 | |
2-chloro-3-nitro-6-methoxypyridine, pyridine, 2-chloro-6-methoxy-3-nitro, 2-chloro-6-methoxy-3-nitro-pyridine, 2-chloro-6-methoxy-3 or 5-nitropyridine, pubchem4044, acmc-1agmz, ksc226c7t, 2-chloro-3nitro-6-methoxy-pyridine, 6-methoxy-2-chloro-3-nitropyridine, 2-chloro-6-methoxy-3-nitropyridine | |
2-chloro-6-methoxy-3-nitropyridine |
Specifications
38533-61-8 | |
White to Yellow | |
MFCD00130268 | |
1454907 | |
Slightly soluble in water. | |
COC1=CC=C(C(Cl)=N1)[N+]([O-])=O | |
188.57 | |
188.57 | |
Powder |
78°C to 80°C (literature) | |
C6H5ClN2O3 | |
UN2811 | |
2-chloro-3-nitro-6-methoxypyridine, pyridine, 2-chloro-6-methoxy-3-nitro, 2-chloro-6-methoxy-3-nitro-pyridine, 2-chloro-6-methoxy-3 or 5-nitropyridine, pubchem4044, acmc-1agmz, ksc226c7t, 2-chloro-3nitro-6-methoxy-pyridine, 6-methoxy-2-chloro-3-nitropyridine, 2-chloro-6-methoxy-3-nitropyridine | |
DVRGUTNVDGIKTP-UHFFFAOYSA-N | |
2-chloro-6-methoxy-3-nitropyridine | |
2795029 | |
98% | |
2-Chloro-6-methoxy-3-nitropyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 253-989-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only