missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-6-methylpyrimidine-4,5-diamine, ≥95%, Thermo Scientific™ Product Code.: 10682453

2-Chloro-6-methylpyrimidine-4,5-diamine, ≥95%, Thermo Scientific™

Product Code. 10682453
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10682453

Brand: Thermo Scientific Maybridge RJC03992CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 63211-98-3
Molecular Formula C5H7ClN4
Molecular Weight (g/mol) 158.59
MDL Number MFCD01764685
InChI Key KSGKEVCHVYUUDN-UHFFFAOYSA-N
Synonym 2-chloro-4,5-diamino-6-methylpyrimidine, 4,5-diamino-2-chloro-6-methylpyrimidine, 2-chloro-6-methyl-pyrimidine-4,5-diamine, 2-chloranyl-6-methyl-pyrimidine-4,5-diamineShow MoreShow Less
PubChem CID 275295
IUPAC Name 2-chloro-6-methylpyrimidine-4,5-diamine
SMILES CC1=NC(Cl)=NC(N)=C1N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.59
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-methylpyrimidine-4, 5-diamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-6-methylpyrimidine-4,5-diamine, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.