missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-6-methylpyrimidine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 10802271
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89581-58-8
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00833002
InChI Key FBXGLOGQWMGIER-UHFFFAOYSA-N
Synonym 2-chloro-6-methyl-pyrimidine-4-carboxylic acid, 2-chloro-6-methyl-4-pyrimidinecarboxylic acid, 2-chloro-4-methylpyrimidine-6-carboxylic acid, 4-pyrimidinecarboxylic acid, 2-chloro-6-methyl
PubChem CID 10535213
IUPAC Name 2-chloro-6-methylpyrimidine-4-carboxylic acid
SMILES CC1=NC(=NC(=C1)C(=O)O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 172°C to 173°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.57
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-methylpyrimidine-4-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-6-methylpyrimidine-4-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-chloro-6-methylpyrimidine-4-carboxylic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.