missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trichloromethyl)pyridine, 98%

Product Code. 15422749
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15422749

Brand: Thermo Scientific Alfa Aesar H66418.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1929-82-4 | C6H3Cl4N | 230.90 g/mol

2-Chloro-6-(trichloromethyl)pyridine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-(trichloromethyl)pyridine is

Solubility
Soluble in 10 mg/ml ethanol.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1929-82-4
Molecular Formula C6H3Cl4N
Molecular Weight (g/mol) 230.90
MDL Number MFCD00072500
InChI Key DCUJJWWUNKIJPH-UHFFFAOYSA-N
Synonym 2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso
PubChem CID 16004
ChEBI CHEBI:81935
IUPAC Name 2-chloro-6-(trichloromethyl)pyridine
SMILES ClC1=CC=CC(=N1)C(Cl)(Cl)Cl

Specifications

Melting Point 60°C to 65°C
Color White
Flash Point 100°C (212°F)
Quantity 1 g
UN Number UN3077
Beilstein 1618997
Solubility Information Soluble in 10mg/ml ethanol.
Formula Weight 230.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-6-(trichloromethyl)pyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-6-(trichloromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Nitrapyrin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.