missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 97%, Thermo Scientific Chemicals

Product Code. 15451096
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1013916-37-4
Molecular Formula C13H14ClN3O
Molecular Weight (g/mol) 263.73
MDL Number MFCD13181207
InChI Key BSKNQSYIDZUXQT-UHFFFAOYSA-N
PubChem CID 44248248
IUPAC Name 2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
SMILES CC1=CC(=O)N(C2CCCC2)C2=NC(Cl)=NC=C12

Specifications

Quantity 5 g
Beilstein 15642863
Synonym 2-chloro-8-cyclopentyl-5-methylpyrido 2,3-d pyrimidin-7 8h-one, 2-chloro-8-cyclopentyl-5-methyl-8h-pyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methylpyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methyl-8h-pyrido 2,3-d py, pyrido 2,3-d pyrimidin-7 8h-one,2-chloro-8-cyclopentyl-5-methyl, pyrido 2,3-d pyrimidin-7 8h-one, 2-chloro-8-cyclopentyl-5-methyl
Solubility Information Slightly soluble in water.
Formula Weight 263.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'