missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-N-(4-iodophenyl)acetamide, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Specifications
Specifications
CAS | 2564-00-3 |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 194.8°C to 196.7°C |
Color | White to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Molecular Formula | C8H7ClINO |
Quantity | 5 g |
Synonym | 2-chloro-n-4-iodophenyl acetamide, n1-4-iodophenyl-2-chloroacetamide, acetamide,2-chloro-n-4-iodophenyl, 4'-iodo-2-chloroacetoanilide, 2-chloro-n-4-iodo-phenyl-acetamide, acetamide, 2-chloro-n-4-iodophenyl, 10.14272/wiuvqcirraapax-uhfffaoysa-n, doi:10.14272/wiuvqcirraapax-uhfffaoysa-n |
Show More |
Safety and Handling
Product Identifier
- 2-Chloro-N-(4-iodophenyl)acetamide
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction