missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-ethylnicotinamide, 97%, Thermo Scientific™

Product Code. 15493968
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52943-22-3
Molecular Formula C8H9ClN2O
Molecular Weight (g/mol) 184.623
MDL Number MFCD01358076
InChI Key LAGOHZCKMJZGIY-UHFFFAOYSA-N
Synonym 2-chloro-n-ethylnicotinamide, n-ethyl-2-chloropyridine-3-carboxamide, 2-chloro 3-pyridyl-n-ethylcarboxamide
PubChem CID 9182570
IUPAC Name 2-chloro-N-ethylpyridine-3-carboxamide
SMILES CCNC(=O)C1=C(N=CC=C1)Cl

Specifications

Quantity 250 mg
Formula Weight 184.63
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-ethylnicotinamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'